Digi-Sense Thermocouple Flexible Wire Probes - FEP-Insulated with Sealed Junction and minconnector
FEP-Insulated with Sealed Junction Used for Long-Term Immersion in Semi-Caustic Environments
  • Each Probe is hermetically sealed and tested to 25psi
  • Can be used for applications requiring an isolated junction.
  • Response Time: 0.75 seconds or greater
  • All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
  • Custom designs or changes to this product are available upon request
Do not subject type J exposed-junction probes to moisture as they will rust
8,200.00 - ₹10,900.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
400 K
₹8,200.00INR / Each
COMPARE
400 J
₹8,200.00INR / Each
COMPARE
400 T
₹8,200.00INR / Each
COMPARE
400 K
₹9,600.00INR / Each
COMPARE
400 T
₹9,600.00INR / Each
COMPARE
400 K
₹10,900.00INR / Each
COMPARE
400 J
₹10,900.00INR / Each
COMPARE
400 T
₹10,900.00INR / Each
Showing 1- 8 of 8