Digi-Sense Thermocouple Bolt-On Stainless Steel Surface Probes with Armored Cable and Stripped Leads
Heavy Duty Bolt-On Probes for Long Term Installation
  • Each Probe comes with a bolt on washer that is .62” OD and .28” ID for quick easy installation
  • Once installed the rugged design allows for continuous long term readings
  • This probe is not recommended for damp or wet conditions
  • Response Time: 3 seconds or greater
  • Each probe comes with a color coded ANSI stripped leads: type J-red/white, type K-red/yellow, and type T-red/blue
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
6,600.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Probe Type
Availability Pricing
COMPARE
900 32 J
₹6,600.00INR / Each