This product is now discontinued, View Related Products
Digi-Sense Silicone Filled Bottom-Connected Dual-Scale Industrial Bimetal Thermometers
Silicone-filled gauges provides for readability and long life in tough industrial environments.
  • Dampened movement
  • Anti-parallax dial for accurate readings
  • Hermetically sealed per ASME B40.3 standard to prevent fogging and protect against tough environments
  • Rugged 304 stainless steel construction with Acrylic lens
  • Recalibrate using the external reset screw
  • For use in applications that do not exceeding 200F
Accuracy is ±1% full-scale when stem is immersed 2" in liquids or 4" in gases (immerse models with a 2.5" stem length 2" in gases). Measured stem length from the top of the threads to the bottom tip of the stem. Stem dia is 0.25"; threaded connection is 1/2" NPT(M). All thermometers meet or exceed ASME B40.3 standards. Maximum environmental temperature for Acrylic lens thermometers is 200degF (93degC).
Discounts: Save 10% on 10-24 assorted thermometers, 15% on 25 to 49, and 20% on 50 or more.
Call For Price
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Dial Size (in) Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
3 250 0
Call For Price
COMPARE
3 300 50
Call For Price
COMPARE
3 250 0
Call For Price
COMPARE
3 300 50
Call For Price
COMPARE
3 250 0
Call For Price
COMPARE
3 300 50
Call For Price
COMPARE
3 250 0
Call For Price
COMPARE
3 300 50
Call For Price
COMPARE
3 250 0
Call For Price
COMPARE
3 300 50
Call For Price
COMPARE
3 250 0
Call For Price
COMPARE
3 300 50
Call For Price
COMPARE
5 250 0
Call For Price
COMPARE
5 300 50
Call For Price
COMPARE
5 250 0
Call For Price
COMPARE
5 300 50
Call For Price
COMPARE
5 250 0
Call For Price
COMPARE
5 300 50
Call For Price
COMPARE
5 250 0
Call For Price
COMPARE
5 300 50
Call For Price
Showing 1 - 20 of 24