Cole-Parmer® Reusable Temperature Indicating Strips
Each style covers a wide range of temperatures, for use in many different application
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.
210.00 - ₹11,100.00INR / Pkg of 10
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max temperature (° F) Temperature Points (° F) Description
Availability Pricing
COMPARE
120 26 to 120°F in 2° increments Reversible 48-Point Vertical Magnetic Temperature Label, 26-120F; 10/Pk
₹11,100.00INR / Pkg of 10
COMPARE
32 32 ,41, 50, 59, 68, 77, 86 Reversible 7-Point Horizontal Temperature Label, -30C-0C/-22-32F; 10/Pk
₹3,000.00INR / Pkg of 10
COMPARE
86 32-86 Reversible 7-Point Horizontal Temperature Label, 0C-32C/32-86F; 10/Pk
₹1,900.00INR / Pkg of 10
COMPARE
140 86-140 Reversible 7-Point Horizontal Temperature Label, 30C-60C/86-140F; 10/Pk
₹1,800.00INR / Pkg of 10
COMPARE
184 140-194 Reversible 7-Point Horizontal Temperature Label, 60C-90C/140-194F; 10/Pk
₹1,600.00INR / Pkg of 10
COMPARE
248 194, 203, 212, 221, 230, 239, 248 Reversible 7-Point Horizontal Temperature Label, 90C-120C/194-248F; 10/Pk
₹1,900.00INR / Pkg of 10
COMPARE
56 26 to 56 Reversible 16-Point Vertical Temperature Label, -3-13C/26-56F; 10/Pk
₹2,100.00INR / Pkg of 10
COMPARE
88 58 to 88 Reversible 16-Point Vertical Temperature Label, 14-31C/58-88F; 10/Pk
₹2,100.00INR / Pkg of 10
COMPARE
120 90 to 120 Reversible 16-Point Vertical Temperature Label, 32-49C/90-120F; 10/Pk
₹2,100.00INR / Pkg of 10
COMPARE
152 122, 124,126, 128, 130, 132, 134, 136, 138, 140, 142, 144, 146, 148, 150, 152 Reversible 16-Point Vertical Temperature Label, 50-66C/122-152F; 10/Pk
₹2,100.00INR / Pkg of 10
COMPARE
Reversible 16-Point Vertical Temperature Label, 68-83C/154-184F; 10/Pk
₹2,100.00INR / Pkg of 10
COMPARE
184 154, 156, 158, 160, 162, 164, 166, 168, 170, 172, 174, 176, 178, 180, 182, 184 Reversible 48-Point Vertical Adhesive Temperature Label, 154-184F; 25/Pk
₹5,200.00INR / Pkg of 25
COMPARE
120 106, 108, 110, 112, 114, 115, 118, 120 Reversible 8-Point Vertical Temperature Label, 41-49C/106-120F; 10/Pk
₹2,700.00INR / Pkg of 10
COMPARE
₹210.00INR / Each
Showing 1- 14 of 14