Cole-Parmer® Reusable Low-Temperature Indicating Strips
Ideal for use in refrigerator, freezer and cooler applications
  • Nontoxic nonhazardous temperature-measuring alternative
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring low temperatures. Indicating strips use liquid crystal technology to display the current temperature range at an economical cost. Shelf life: 1 year after date of purchase.
4,300.00 - ₹6,000.00INR / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Max temperature (° F) Temperature Points (° F)
Availability Pricing
COMPARE
5 1/2 54 28 to 54
₹6,000.00INR / Pkg of 10
COMPARE
5 1/2 30 (-5) to 30
₹6,000.00INR / Pkg of 10
COMPARE
5 3/8 50 34 to 50
₹4,300.00INR / Pkg of 10