This product is now discontinued, View Related Products
Digi-Sense™ Receptacle Testers
The fast and effortless way to test the safety of your electrical socket outlets
  • Inspect common faulty wiring issues in main socket installations
  • Easily find faults with bright light indicators
Check the safety of electrical socket outlets by detecting wiring faults and incorrect grounding. Plug the tester into a standard 3-wire outlet, and different light combinations will indicate common wiring faults found in electrical sockets. Select from two types: circuit tester or GFCI circuit breaker tester. For use in standard 110 to 125 VAC outlets.
2,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Circuit Tester
Call For Price
COMPARE
GFCI Tester
₹2,000.00INR / Each