Digi-Sense Precalibrated Professional Thermocouple Thermometers
Rugged, heavy-duty construction protects against knocks and jars
  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards
  • Relative time stamp provides a time reference for major events
  • Electronic off set function allows compensation of probe errors
  • Infrared input port allows safe, noncontact measurements with optional IR wand
These professional type K thermometers provideaccurate and... reliable temperature measurement in an ergonomic design. Double-molded heavy-duty housing provides superior protection, making them ideal for tough applications. Plus the NIST-traceable precalibration means you get reliable measurements right out of the box, saving you time and money!
All models feature Min/Max/Avg, Hold, °F/°C or Kelvin (K) selectable units, relative time stamp on Min/Max/Avg, electronic offset function, temperature differential, backlit display, and auto power-off. Accept a wide variety of type K thermocouple probes with miniconnector.
These meters also feature an infrared input that accepts an optional IR wand for noncontact measurements of objects that are difficult or dangerous to reach; order wand 20250-24 separately below. The IR temperature reading can be used as a unique input when calculating temperature differential. The IR wand has a distance-to sight ratio of 8:1 and temperature range of –31 to 1022°F (–35 to 550°C).
45,400.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Data Logging Number Of Probe Channels Types Of Probes Accepted
Availability Pricing
COMPARE
Yes 4 Type K
₹45,400.00INR / Each