Traceable® Precalibrated Professional Infrared Thermometers
Safely and easily locate hot and cold spots, preventing problems and saving costs
  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards
  • High distance-to-sight (D/S) ratios handle your long-distance measurements
  • External type K input for contact measurements
  • Dual-laser sighting to pinpoint target areas
  • Protective lens cover with magnetic lock
  • Pistol-grip handle is comfortable and easy to use
  • Measure surface temperatures quickly and accurately
  • USB interface for easy data transmission
These professional IR thermometers are specifically designed for demanding applications with a high distance-to-target ratio, enhanced temperature range, and heavy-duty design. Plus the NIST-traceable precalibration means you get reliable measurements right out of the box, saving you time and money!
Most IR thermometers require the user to be to within 8 to 12 inches or even less to get an accurate reading. Dual targeting lasers give a more precise indication of the measurement area; unlike IR thermometers with a single targeting laser where the user has no idea how big of a spot is being measured.
Both models feature Min/Max/Avg and Hold readings, temperature differential, high/low alarms, °F/°C selectable units, adjustable emissivity, trigger lock for scanning, large backlight LCD, USB interface, and auto power-off.
53,500.00 - ₹62,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Emissivity Max Temperature (° F)
Availability Pricing
COMPARE
30:1 Adjustable 3002
₹53,500.00INR / Each
COMPARE
50:1 Adjustable 3362
₹62,100.00INR / Each