Digi-Sense Magnet Mount Surface Bimetal Thermometers
Inexpensive way to easily measure surface temperature
  • For reference only, not intended for applications where a precise reading is required
  • Item is not recommended to be calibrated
An ideal choice for use on ovens, boilers, process lines, motors, or anywhere a surface temperature needs to be measured. The specially designed bimetal sensing element and housing provide quick readings. These thermometers feature a dual magnet mount that will attach to an ferrous surface, steel case, glass lens and polished aluminum 2" dial with easy-to-read graduations.
6,000.00 - ₹12,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Dial Size (in) Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
2 500 0
₹12,200.00INR / Each
COMPARE
2 750 50
₹6,000.00INR / Each