Cole-Parmer® Irreversible Low-Temperature Indicators / Descending Labels
Reliable proof that the product was maintained at the correct temperature
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
These labels offer a highly reliable and irreversible indication of exposure to low temperatures. Labels clearly indicate when the product to which it is attached to has been subjected to harsh temperatures, which could have detrimental effects on refrigerated pharmaceuticals, foods, chemicals, paints, and adhesives. Available in both ascending and descending style Accuracy is ±1°C of the rated temperature. Shelf life: one year after date of purchase.
12,200.00INR / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Number of Temperature Indicators Temperature Points (° F)
Availability Pricing
COMPARE
1/4 1 32
₹12,200.00INR / Pkg of 10
COMPARE
1/4 1 36
₹12,200.00INR / Pkg of 10