Cole-Parmer® Irreversible High-Temperature 6-Point Horizontal Labels / Indicators
Reliable proof that the product was maintained at the correct temperature
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8°F below 212°F and ±1°F above 212°F. Shelf life: 1 year after date of purchase.
3,700.00 - ₹4,200.00INR / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Max temperature (° F) Temperature Points (° F)
Availability Pricing
COMPARE
1 1/4 108 84, 93, 99, 104, 108, 111
₹3,700.00INR / Pkg of 10
COMPARE
1 1/4 144 111, 115, 120, 129, 140, 144
₹4,200.00INR / Pkg of 10
COMPARE
1 1/4 199 149, 160, 171, 180, 190, 199
₹3,700.00INR / Pkg of 10
COMPARE
1 1/4 261 210, 219, 230, 241, 250, 261
₹3,700.00INR / Pkg of 10
COMPARE
1 1/4 320 270, 280, 289, 300, 309, 320
₹3,700.00INR / Pkg of 10
COMPARE
1 1/4 379 331, 340, 351, 360, 370, 379
₹3,700.00INR / Pkg of 10
COMPARE
1 1/4 450 390, 399, 410, 421, 435, 450
₹3,700.00INR / Pkg of 10
COMPARE
1 1/4 554 460, 480, 489, 500, 536, 554
₹3,700.00INR / Pkg of 10
Showing 1- 8 of 8