Digi-Sense Food Safety IR Thermometer with Probe
A combination contact/noncontact thermometer for all food applications
  • Go/no-go LEDs provide quick check of HACCP food zones
  • Sealed IP54 housing make for easy cleaning
Check critical food temperatures and monitor HACCP food safety zones with this combination contact/noncontact thermometer. LED’s rapidly indicate if food temperature is in the bacterial growth safety zone. Green lights indicate safe temperatures below 40°F (4°C) and above 140°F (60°C). Red light indicates the HACCP danger zone between 40 to 140°F (4 to 60°C). The contact probe swings out for internal temperature checks, then folds in for storage. IR mode provides quick, noncontact surface measurements and stores maximum reading. Target illumination indicates target measurement area and is ideal for close working distances of 2 to 12”. Countdown timer with alarm monitors cooking and cooling intervals and HACCP exposure times.
33,900.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Distance To Target Size Ratio (D:S) Max Temperature (° F) Description
Availability Pricing
COMPARE
2.5:1 525 Food Safety Infrared (IR) Thermometer with Probe
₹33,900.00INR / Each