Digi-Sense® Digital Pocket Thermometers
Quickly verify termperature
These thermometers are ideal for laboratory, field, and process measurements. Use to quickly verify termperature. Typical response time is 30 seconds for 99% of new reading. Thermometers have a 304 stainless steel penetration stem. All models include a protective case with pocket clip and a battery. Thermometer features a sealed case—ideal for use with liquids. It has °>F/°>C switchable ranges, a MIN/MAX button, and a one-second display update. Use ON/OFF button to conserve battery life.
5,300.00 - ₹10,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
COMPARE
392 14 Water-Resistant Digital Pocket Thermometer; 5\"L x 0.14\"OD Probe
₹10,000.00INR / Each
COMPARE
302 -58 Pocket Thermometer; Range -58 to 302°F (-50 to 150°C)
₹5,700.00INR / Each
COMPARE
302 -58 Long-Stem Digital Pocket Thermometer, 8\" L, -58 to 302F/-50 to 150C
₹5,300.00INR / Each