This product is now discontinued, View Related Products
Digi-Sense Dampened Adjustable Angle Dual-Scale Industrial Bimetal Thermometers
The benefits of a liquid-filled case in a silicone-free dry configuration
  • Dampened movement
  • More economical than silicone gauges
  • Greater temperature range than silicone-filled gauges
  • Anti-parallax dial for accurate readings
  • Eliminate the risk of a leaking case and cross-contamination
  • Hermetically sealed per ASME B40.3 standard to prevent fogging and protect against tough environments
  • Rugged 304 stainless steel construction with a clear glass lens
  • Recalibrate using the external reset screw
Accuracy is ±1% full-scale when stem is immersed 2" in liquids or 4" in gases (immerse models with a 2.5" stem length 2" in gases).
Measured stem length from the top of the threads to the bottom tip of the stem. Stem dia is 0.25"; threaded connection is 1/2" NPT(M). All thermometers meet or exceed ASME B40.3 standards.
Maximum environmental temperature for glass lens thermometers is 500degF (148degC).
Discounts: Save 10% on 10-24 assorted thermometers, 15% on 25 to 49, and 20% on 50 or more.
Call For Price
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Dial Size (in) Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
3 250 0
Call For Price
COMPARE
3 550 50
Call For Price
COMPARE
3 250 0
Call For Price
COMPARE
3 550 50
Call For Price
COMPARE
3 250 0
Call For Price
COMPARE
3 550 50
Call For Price
COMPARE
3 250 0
Call For Price
COMPARE
3 550 50
Call For Price
COMPARE
3 250 0
Call For Price
COMPARE
3 550 50
Call For Price
COMPARE
3 250 0
Call For Price
COMPARE
3 550 50
Call For Price
COMPARE
5 250 0
Call For Price
COMPARE
5 550 50
Call For Price
COMPARE
5 250 0
Call For Price
COMPARE
5 550 50
Call For Price
COMPARE
5 250 0
Call For Price
COMPARE
5 550 50
Call For Price
COMPARE
5 250 0
Call For Price
COMPARE
5 550 50
Call For Price
Showing 1 - 20 of 24