Digi-Sense® Blue Spirit Pocket Glass Thermometers, Metal Case, Celsius-scale
Low-cost way to measure temperature with a compact and durable metal case
  • Non-mercury blue spirit liquid
These thermometers are ideal for use in laboratory, biology, chemistry, university, photographic, food/beverage, petrochemical, automotive, wastewater and many other general purpose applications. The protective plastic case protects these thermometers from accidental breakage during use and storage. The section of the case that surrounds the bulb has small openings for circulation. All DURAC Plus® thermometers feature markings that are permanently printed on the glass, blue spirit liquid that has a dark scale contrasts against a white background.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
COMPARE
50 -35 160
Call For Price
COMPARE
110 -10 160
Call For Price
COMPARE
50 -5 160