Digi-Sense® Benchtop Temperature Control Consoles
Direct plug-in of heating or cooling devices
  • Simple on/off control to sophisticated auto-tuning PID control
  • Simultaneous view measured value and set point on two-line display
  • Front panel indicates output and alarm conditions
  • Grid support bracket ideal for fume hood mounting
The benchtop single-loop temperature controllers are designed to regulate a user-defined set point temperature. The microprocessor control provides heating/cooling tolerances for demanding applications found in laboratory or industrial environments. Temperature scale selectable to read in °F, °C, K (Kelvin), or °R (Rankine). An auto-tuning function provides automatic configuration of the PID parameters with minimum user configuration required. Nonvolatile memory stores all user-defined information.
A convenient output receptacle, located on the back of the units, allows for direct plug-in of heaters and other devices. All models accept eight thermocouple types (J, K,N,R,S,T B, and E). Safety features include a loop-break alarm, overtemperature and timer cutoffs, separate fuses for instrument and output receptacle, and audible/visible out-of-range alarms.
222,000.00 - ₹288,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Power (VAC) Description
Availability Pricing
COMPARE
2502 120 Benchtop Temperature Controller for Thermocouples; 120 VAC
₹222,000.00INR / Each
COMPARE
2502 230 Benchtop Temperature Controller for Thermocouples; 230 VAC
₹222,000.00INR / Each
COMPARE
2502 120 Benchtop Temperature Controller with USB Output, for Thermocouple, RTD, or Thermistors; 120 VAC
₹288,000.00INR / Each
COMPARE
2502 230 Benchtop Temperature Controller with USB Output, for Thermocouple, RTD, or Thermistors; 230 VAC
₹288,000.00INR / Each