Digi-Sense AM1660 Series High-Temperature Precision Industrial PRT/RTD Probes
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
AM1660 Series-High temperature precision industrial PRT/RTD probes: This is a high temperature version of the AM1640 probe. Manufactured using a coil suspension element design for increased shock and vibration resistancealong with flexibility and durability. The AM1660 has a temperature range of -200 to 670ºC (-328 to 1238ºF).
Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.
216,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
Stripped Wire 1238 -328
₹216,000.00INR / Each