This product is now discontinued, View Related Products
Digi-Sense AM1643 Series Full Immersion PRT / RTD Probes
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • Equivalent to Fluke Calibration / Hart Scientific Model 5607
AM1612 and AM1643 Series—Full-immersion PRT/RTD probes: Designed to perform in extreme environments where both the transition junction and the lead wires are required to withstand temperatures covering the entire operating range of the probe. Applications include calibration or validation of sensors used in laboratory or biology freezers, walk-in refrigerators, autoclaves, ovens, stability test chambers, furnaces, and incubators. These probes feature short term stability of 20 mK at TPW and long term drift of 40 mK at Triple Point of Water (TPW). The AM1643 has a range of 0 to 420ºC (32 to 788ºF).
Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.
194,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Connector Type Max Temperature (° F)
Availability Pricing
COMPARE
Stripped Wire 788
Call For Price
COMPARE
Spade Lugs 788
₹194,000.00INR / Each
COMPARE
Mini Spade Lug 788
Call For Price
COMPARE
5-Pin DIN 788