This product is now discontinued, View Related Products
Digi-Sense AM1640 Series-Precision Industrial PRT / RTD Probes
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • Equivalent to Fluke Calibration / Hart Scientific Model 5627A
  • .
AM1640 Series—Precision industrial PRT/RTD probes: Conform to the standard 385 curve, letting you use your DIN/IEC RTD meters fully. Manufactured using a coil suspension element design for increased shock and vibration resistancealong with flexibility and durability. All have an accuracy of 35 mK at 0°C and short-term stability of 10 mK at 0°C. The AM1640 has a range of -200 to 420ºC (-328 to 788ºF).
Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Connector Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
Stripped Wire 788 -328
Call For Price
COMPARE
Spade Lugs 788 -328
Call For Price
COMPARE
Mini Spade Lug 788 -328
Call For Price
COMPARE
5-Pin DIN 788 -328