Digi-Sense AM1620 Series-Precision Industrial PRT / RTD Probes
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
AM1620 Series—Precision industrial PRT/RTD probes: Conform to the standard 385 curve, letting you use your DIN/IEC RTD meters fully. Manufactured using a coil suspension element design for increased shock and vibration resistancealong with flexibility and durability. All have an accuracy of 35 mK at 0°C and short-term stability of 10 mK at 0°C. The AM1620 has a range of -60 to 300ºC (-76 to 572ºF)
Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.
107,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
Stripped Wire 572 -76
₹107,000.00INR / Each