Digi-Sense 1.75" Dial Stainless Steel Pocket Thermometers
Choose the dial size and temperature range to meet your specific needs
  • Factory tested to three points with a ±1% accuracy
  • Minimum immersion: liquids 2" (50 mm), gases 4" (100 mm)
  • Hermetically sealed dial stops internal fogging
  • Recalibrate using external adjustment nut
  • 304 Stainless Steel dial case and stem
  • Shock resistant
2,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Dial Size (in) Stem Length (in) Description
Availability Pricing
COMPARE
1.75 8 S.S. Bimetalal Pocket Thermo -100-40C, 1.75\" Dial, Glass Lens, 8\" Stem, 2F Div
₹2,500.00INR / Each
COMPARE
1.75 8 S.S. Bimetalal Pocket Thermo -40-160F, 1.75\" Dial, Glass Lens, 8\" Stem, 2F Div
₹2,500.00INR / Each