Dickson Circular Chart Recorder Accessories

Find probes, cables, and more!
  • Temperature probes
  • Software
  • Communication cables
  • AC adapter
For use with Dickson brand chart recorders.
8,900.00 - ₹36,500.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Dickson D605 4" Piercing Probe for Dual-Pen Datalogging Temperature Recorder WW-08516-03 Mfr # D605
COMPARE
4\" Piercing Probe for Dual-Pen Datalogging Temperature Recorder
₹29,500.00INR / Each
Dickson A203 6" High Temperature Probe for Dual-Pen Datalogging Temperature Recorder WW-08516-06 Mfr # A203
COMPARE
6\" High Temperature Probe for Dual-Pen Datalogging Temperature Recorder
₹36,500.00INR / Each
Dickson A792 Steam Pressure Pigtail for PW4/PW8 Chart Recorders WW-68511-61 Mfr # A792
COMPARE
Steam Pressure Pigtail for PW4/PW8 Chart Recorders
₹11,200.00INR / Each
Dickson A015 Data Logger Software and 9-Pin Serial Cable WW-80009-50 Mfr # A015
COMPARE
Data Logger Software and 9-Pin Serial Cable
₹19,400.00INR / Each
Dickson R065 Universal International AC Adapter, 100 to 240 VAC WW-80018-20 Mfr # R065
COMPARE
Universal International AC Adapter, 100 to 240 VAC
₹8,900.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat