This product is now discontinued, View Related Products
Dickson Large Display Temperature and Humidity Data Loggers
Large 1-3/8'' tall display can be seen from across the lab — up to 25 feet away!
  • Up to 32,000 datapoints can be stored in memory
These dataloggers feature high/low alarms, min/max readings, delayed start times, and °F/C selections. Download data to your PC via USB, 9-pin serial port, or use the portable FLASH memory card. Software allows for simple viewing and zooming of logged data in a graphical or tabular (numeric table) format, order separately.
Optional 21CFR11 software meets pharmaceutical requirements including, password protection, electronic signatures, encrypted data in secured files and auditable trails to identify users, date & times and actions taken.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Input Type Max Temperature (° F) Temperature Accuracy (° F)
Availability Pricing
Dickson SM300 Temperature Data Logger with Large Display and USB Port WW-23036-00 Mfr # SM300
COMPARE
Temperature (one external) 158 1.8
Call For Price
Dickson SM320 Datalogger, large display, temperature 2-channel WW-23036-10 Mfr # SM320
COMPARE
Temperature (one internal, one external) 2000 1.8
Call For Price
Dickson SM325 Datalogger, large display, temperature 2-channel; Type K WW-23036-20 Mfr # SM325
COMPARE
Temperature (two external) 2000 1.8
Call For Price
Dickson TM320 Datalogger, large display, temperature and humidity WW-23036-30 Mfr # TM320
COMPARE
Temperature and humidity 158 0.8
Call For Price
Dickson TM325 Datalogger, large display, humidity and temperature with remote probe WW-23036-40 Mfr # TM325
COMPARE
Temperature (one external) and humidity 158 0.8
Call For Price
Dickson SM420 LCD display temperature data logger with external platinum RTD probe WW-68511-86 Mfr # SM420
COMPARE
Temperature 350 0.5