This product is now discontinued, View Related Products

Dickson 8-inch Temperature/Humidity Recorders

Simultaneously view temperature and humidity or dew point
  • Selectable units and scales—choose chart paper based on your needs
  • No inadvertent changes with lock-out dip switch
Select your own temperature range and scale; humidity or dew point scale; and 24-hour, 7-day, or 31-day rotation period (Charts available separately) Prevent setting changes with lock-out dip switch. Rear panel features keyhole slots for wall mounting.
These rugged and compact recorders use microprocessor-based replaceable electronic sensors to record temperature and humidity measurements. Units feature a digital display with high/low alarms and a SPST 24 V, 500 mA relay capable of shutting down a process in alarm status.
181,000.00 - ₹216,000.00INR / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Chart Diameter (in) Max Temperature (° C) Description
Availability Pricing
Dickson TH8P2 8" Temperature/Humidity Recorder with Digital Display WW-80001-51 Mfr # TH8P2
COMPARE
8 120 8\" Temperature/humidity Recorder With Digital Display 80001-51
₹181,000.00INR / Each
Dickson TH8P2 8" Temperature/humidity Recorder With Digital Display 80001-52 WW-80001-52 Mfr # TH8P2
COMPARE
8 120 8\" Temperature/humidity Recorder With Digital Display 80001-52
Call For Price
Dickson TH8P3 8" Temperature/Humidity Recorder with Probe, Digital Display And Alarms WW-80001-53 Mfr # TH8P3
COMPARE
8 185 8\" Temperature/humidity Recorder With Probe, Dig. Display And Alarms
₹216,000.00INR / Each
Dickson TH803 8" Temperature/humidity Recorder With Probe, Dig. Display And Alarms WW-80001-54 Mfr # TH803
COMPARE
8 185 8\" Temperature/humidity Recorder With Probe, Dig. Display And Alarms
Call For Price
Dickson TH805 8" Temperature/humidity Recorder With Probe, Dig. Display Alarms, And Relays. WW-80001-55 Mfr # TH805
COMPARE
8 185 8\" Temperature/humidity Recorder With Probe, Dig. Display Alarms, And Relays.
Call For Price
Dickson TH800 Battery-Powered 8" Temperature/Humidity Recorder WW-80001-56 Mfr # TH800
COMPARE
8 120 Battery-Powered 8\" Temperature/Humidity Recorder
Call For Price
Dickson TH8P5 8" Humidity/Temperature Recorder; 120 VAC/Battery WW-80001-57 Mfr # TH8P5
COMPARE
8 185 8\" Humidity/Temperature Recorder; 120 VAC/Battery
Call For Price
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat