Dickson 6” Type K Thermocouple Recorders

Monitor temperature for quality assurance and control
  • Audible alarm notifies operator of out-of-limit conditions
  • Shielded design eliminates electronic noise interference
These microprocessor-based recorders are ideal for monitoring temperatures in ovens, freezers, chambers, storage and clean room facilities, machinery and more. Recorders feature user selectable temperature ranges, recording times, zero point calibration, and include a 4-ft flexible type K thermocouple bead wire. The large digital LCD is easy to read and has a resolution of 0.1°F/°C.
Order charts separately. Units are enclosed in a rugged ABS case with a polycarbonate door to withstand challenging environments and are portable or wall mountable with rear-panel keyhole slots.
116,000.00 - ₹188,000.00INR / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Chart Diameter (in) Chart Speed Number of Channels
Availability Pricing
Dickson KT621 6" Temperature Chart Recorder WW-80018-00 Mfr # KT6P1
COMPARE
6 24 hour; 7 day (user selectable) 1
₹116,000.00INR / Each
Dickson KT622 6" temperature recorder with remote probe WW-80018-02 Mfr # KT6P2
COMPARE
6 24 hour; 7 day (user selectable) 1
₹147,000.00INR / Each
Dickson KT6P5  6" Temperature Chart Recorder WW-80018-06 Mfr # KT6P5
COMPARE
6 24 hour; 7 day (user selectable) 1
₹188,000.00INR / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat