This product is now discontinued, View Related Products
Diba Omnifit® Rotary Valves with Omnifit Cap System
PTFE body and PCTFE rotor offer excellent chemical resistance
  • Pressure rated up to 50 psi (3.4 bar)
Rotary valves have a 1.5 mm bore and accept tubing sizes between 1 and 4 mm OD. Select from various configurations to suit your application.
38,500.00 - ₹54,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description Flow Pattern
Availability Pricing
Diba Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 4-port, 4-way, 90° flow; 1/ea WW-21939-20 Mfr # 001112
COMPARE
Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 4-port, 4-way, 90° flow; 1/ea 4 ports, one adjacent pair at 90°
₹38,500.00INR / Each
Diba Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 4-port, 3-way, tee flow; 1/ea WW-21939-21 Mfr # 001113
COMPARE
Omnifit® Standard Valve for1 to 4 mm OD Tubing, 4-port, 3-way, tee flow; 1/ea 4 ports, three ports in a \"T\"
₹41,000.00INR / Each
Diba Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 4-port, 2-way, adjacent flow; 1/ea WW-21939-22 Mfr # 001114
COMPARE
Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 4-port, 2-way, adjacent flow; 1/ea 4 ports, two adjacent pairs at 90°
₹54,900.00INR / Each
Diba Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 5-port, 5-way, 1 common port; 1/ea WW-21939-23 Mfr # 001118
COMPARE
Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 5-port, 5-way, 1 common port; 1/ea 5 ports, one port to any of other 4 ports
₹41,000.00INR / Each