Diba Omnifit® Multiport PTFE Cap System Fittings and Valves
Connect multiple fluid lines with ease
  • Select valves with on/off valves to control your fluid line
  • Use with 1 to 4 mm OD tubing
Quickly and easily connect semi-rigid tubing such as PTFE, ETFE, and FEP as well as rigid materials such as glass, PEEK, and stainless steel. Use the interchangeable components to create chemically inert fluid pathway connections and flat-bottom ports (using Gripper fittings). Fittings are autoclavable (remove caps first). Order colored caps quickly and easily color code multiple fluid lines.
62,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Number Of Ports Description
Availability Pricing
Diba Omnifit® Standard Connector for 1 to 4 mm OD Tubing, 4-way, cross; 1/ea WW-21939-03 Mfr # 001004
COMPARE
PTFE/PP 4 Omnifit® Standard Connector for 1 to 4 mm OD Tubing, 4-way, cross; 1/ea
Call For Price
Diba Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 8-way; 1/ea WW-21939-15 Mfr # 001103
COMPARE
PTFE/PP 8 Omnifit® Standard Valve for 1 to 4 mm OD Tubing, 8-way; 1/ea
₹62,000.00INR / Each