This product is now discontinued, View Related Products

Desco Metal-In ESD Shielding Bags

Protect important electrical components from static charges
  • Content identification made easy with transparent design
These bags provide a static safe environment for electronic devices. The integral antistatic, low tribocharging properties prevent electrostatic charges during movement. The bags consist of three layers: an inner and outer static dissipative polyester layer, and a middle aluminum-shielding layer. The aluminum layer has the properties to discharge 5,000 volts to 0 volts in 50 µsec. The bags feature an amine-free construction allowing compatibility with polycarbonate for a non-corrosive, noncontaminating environment.

Select from bags with or without zippers. Zippered bags feature a low charging zipper for a convenient, recloseable seal. Bags without zippers are heat sealable. Packs of 100 bags.
Call For Price

22 variations of this product are available.

See All Product Options Below
22 PRODUCT OPTIONS
Compareitem specifications
Accuracy Power (VAC) Range
Availability Pricing
Desco 13405 Metal-In ESD Shielding Bags, 3 x 5", 100/Pk WW-26808-20 Mfr # 13405
COMPARE
Call For Price
Desco 13415 Metal-In ESD Shielding Bags, 4 x 6", 100/Pk WW-26808-22 Mfr # 13415
COMPARE
Call For Price
Desco 13425 Metal-In ESD Shielding Bags, 4 x 30", 100/Pk WW-26808-24 Mfr # 13425
COMPARE
Call For Price
Desco 13430 Metal-In ESD Shielding Bags, 5 x 8", 100/Pk WW-26808-26 Mfr # 13430
COMPARE
Call For Price
Desco 13440 Metal-In ESD Shielding Bags, 6 x 8", 100/Pk WW-26808-28 Mfr # 13440
COMPARE
Call For Price
Desco 13460 Metal-In ESD Shielding Bags, 8 x 10", 100/Pk WW-26808-30 Mfr # 13460
COMPARE
Call For Price
Desco 13465 Metal-In ESD Shielding Bags, 8 x 12", 100/Pk WW-26808-32 Mfr # 13465
COMPARE
Call For Price
Desco 13470 Metal-In ESD Shielding Bags, 10 x 12", 100/Pk WW-26808-34 Mfr # 13470
COMPARE
Call For Price
Desco 13475 Metal-In ESD Shielding Bags, 10 x 14", 100/Pk WW-26808-36 Mfr # 13475
COMPARE
Call For Price
Desco 13500 Metal-In ESD Shielding Bags, 12 x 16", 100/Pk WW-26808-38 Mfr # 13500
COMPARE
Call For Price
Desco 13515 Metal-In ESD Shielding Bags, 15 x 18", 100/Pk WW-26808-40 Mfr # 13515
COMPARE
Call For Price
Desco 13520 Metal-In ESD Shielding Bags, 18 x 18", 100/Pk WW-26808-42 Mfr # 13520
COMPARE
Call For Price
Desco 13524 Metal-In ESD Shielding Bags, 18 x 24", 100/Pk WW-26808-44 Mfr # 13524
COMPARE
Call For Price
Desco 13605 Shielding Bag with Zipper, 3" x 5" WW-26808-50 Mfr # 13605
COMPARE
Call For Price
Desco 13615 Shielding Bag with Zipper, 4" x 6" WW-26808-52 Mfr # 13615
COMPARE
Call For Price
Desco 13630 Shielding Bag with Zipper, 5" x 8" WW-26808-54 Mfr # 13630
COMPARE
Call For Price
Desco 13660 Shielding Bag with Zipper, 8" x 10" WW-26808-56 Mfr # 13660
COMPARE
Call For Price
Desco 13665 Shielding Bag with Zipper, 8" x 12" WW-26808-58 Mfr # 13665
COMPARE
Call For Price
Desco 13670 Shielding Bag with Zipper, 10" x 12" WW-26808-60 Mfr # 13670
COMPARE
Call For Price
Desco 13675 Shielding Bag with Zipper, 10" x 14" WW-26808-62 Mfr # 13675
COMPARE
Call For Price
Showing 1 - 20 of 22

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat