Delmhorst TechScan and ProScan Nondestructive Moisture Meter
Trace the source of moisture without destroying your sample
  • Pinless measurement up to 3⁄4" (1.9 cm) deep
  • Scan wood, concrete, and other building materials
Trace the source of moisture intrusion in wood, concrete, and other materials — ideal for contractors, inspectors, and laboratory professionals. Meter measures moisture content and shows the results on the easy-to-read display. Includes an internal calibration check, adjustable alarm, HOLD function, and a color-coded reference scale for nonwood material. Rugged electrode pads measure moisture penetration up to 3/4" deep without destroying the sample. Sensor measuring area is 2-1/2" x 3-1/2".
112,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Measurement Type
Availability Pricing
Delmhorst ProScan Proscan Digital Non-Destructive Moisture Meter WW-59820-91 Mfr # PROSCANW/CS
COMPARE
1 9V (included) Pinless
₹112,000.00INR / Each