Delmhorst Navigator Moisture Meters
Quickly and easily locate water damaged areas by checking moisture content
  • Ensure structures are being properly dried
  • Provides accurate and verifiable moisture content readings
  • Two measuring modes: built-in pins, or nondestructive pinless scanner
  • Expand capabilities with optional electrodes
  • Designed specifically for the Restoration Contractors
These versatile restoration meters offer a moisture mapping system that allows you to easily set up as many as five individual jobs and 32 locations per job. Within each job, designate which rooms have been affected by water damage and specify the material for the floor, ceiling and wall in each room. Take basic readings or set up job definitions by selecting a wood or non-wood type, temperature of material, and adjustable alarm status. Store up to 1400 readings and view statistics for each job via optional software. Display the stored readings or show the average, standard deviation, minimum, and maximum values. Date/time stamp on all stored readings helps with quality control programs.
Navigator Meter combines pin and scan modes to measure moisture content in drywall, wood and other building materials. The pin mode is used to locate the exact source of moisture. The pin mode reads moisture in wood and has a drywall scale that gives actual moisture content. Scan(pinless) mode provides a numerical reference scale that allows you to quickly scan large areas and to identify if a general problem exists. Navigator can also be paired with a variety of electrodes, giving you the flexibility to measure drywall, flooring, insulation, and other materials.
NavigatorPro is a 3-in-1 meter that offers all the same features and specifications as Navigator plus adds on a thermohygrometer to measure temperature and humidity and calculates dew point, and GPP (Grains Per Pound).
300,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Measurement Type
Availability Pricing
Delmhorst NavPro/BP Navigatorpro Moisture Meter, Basic Package WW-59820-67 Mfr # NavPro/BP
COMPARE
1 9V (included) Combination
₹300,000.00INR / Each