Delmhorst J-Series Moisture Meters for Wood
The J-series moisture meters are a great value for measuring moisture content in wood
  • Measures through built-in pins and optional pin electrodes
This popular meter uses a wood scale that ranges from 6 to 40% with built-in calibrations for 48 individual wood species. The contact pins mounted on top of the meter provide 5/16'' (0.8 cm) penetration for testing wood up to 1” (2.5 cm) thick. The meter also features an audible out of range alarm, internal calibration check, 100 data point memory, and average/maximum readings.
48,900.00 - ₹135,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Measurement Type
Availability Pricing
Delmhorst J-LITE Wood Moisture Meter 6-30% WW-59820-00 Mfr # J-LITE
COMPARE
1 9V (included) Pin
₹48,900.00INR / Each
Delmhorst J-2000 Digital Pin-Type Wood Moisture Meter, Basic Package WW-59820-04 Mfr # J-2000
COMPARE
1 9V (included) Pin
₹97,800.00INR / Each
Delmhorst J-2000/PKG Digital Pin-Type Wood Moisture Meter, Expanded Package WW-59820-49 Mfr # J-2000/PKG
COMPARE
1 9V (included) Pin
₹135,000.00INR / Each