Delmhorst BD-2100 Digital Moisture Meters
Ideal for testing building materials such as wood, plaster, concrete, roofing, and insulation
  • Measures through built-in pins expand measuring capability with optional pin electrodes
  • This best selling moisture meter in the industry for years because of its versatility
The BD-2100 meter offers three separate scales: wood, nonwood, and drywall. Built-in colored LED (green/yellow/red) gives you a quick visual indication of where the reading is within each given scale. The wood scale measure moisture in materials such as flooring, studs, joists, and exterior sheathing over the range of 6 to 40%; the nonwood (reference) scale provides comparative readings in materials such as concrete, plaster and insulation; and drywall scale measures actual moisture content within drywall. This easy-to-use meter also features an audible out of range alarm, internal calibration check, 100 data point memory, and average/maximum readings.
See complete offering of optional and replacement electrodes and accessories
103,000.00 - ₹146,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Measurement Type
Availability Pricing
Delmhorst BD-2100 Digital Pin-Type Moisture Meter, Basic Package WW-59820-06 Mfr # BD-2100
COMPARE
1 9V (included) Pin
₹103,000.00INR / Each
Delmhorst BD-2100/EIFS/PK Digital Pin-Type Moisture Meter, EIFS Package WW-59820-07 Mfr # BD-2100/EIFS/PK
COMPARE
1 9V (included) Pin
₹146,000.00INR / Each
Delmhorst BD-2100/PKG Digital Pin-Type Moisture Meterinsulation Package WW-59820-08 Mfr # BD-2100/PKG
COMPARE
1 9V (included) Pin
₹133,000.00INR / Each
Delmhorst BD-2100/26/PKG Digital Pin-Type Moisture Meter, Flooring Package WW-59820-09 Mfr # BD-2100/26/PKG
COMPARE
1 9V (included) Pin
₹139,000.00INR / Each