Defelsko® Surface Testing Accessories

Optimize surface testing
  • Calibration block
  • Ultrasonic gel
Ultrasonic couplant and calibration step blocks ensure accurate and reliable readings.
53,800.00 - ₹120,000.00INR / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
DeFelsko B1 Accessory Block(Inches) 0.1 0.2 0.3 0.4 0.5 WW-59784-27 Mfr # STDB1
COMPARE
Accessory Block(Inches) 0.1 0.2 0.3 0.4 0.5
₹107,000.00INR / Each
DeFelsko B1M Block Metric 2.5 5.0 7.5 10.0 12.5MM WW-59784-28 Mfr # STDB1M
COMPARE
Block Metric 2.5 5.0 7.5 10.0 12.5MM
₹120,000.00INR / Each
DeFelsko GEL P Accessory Case Of 12 Ultrasonic Gel WW-59784-29 Mfr # GEL P
COMPARE
Accessory Case Of 12 Ultrasonic Gel
₹53,800.00INR / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat