Defelsko PosiTector Standard Ultrasonic Coating Thickness Gauges for Non-Metals
Quickly measure coating thickness on all non-metal substrates
  • Ready to measure - no calibration adjustment required to measure most coatings
  • Solvent, acid, oil, water and dust resistant - meets or exceeds IP5X
  • Internal memory for 250 readings in a single batch to be view or downloaded
  • Statistics mode continuously displays average, standard deviation, and number of readings while measuring
  • High contrast backlit display for bright or dark environments
  • Certificate of calibration showing traceability to NIST or PTB included (Long Form)
  • Proven non-destructive ultrasonic technique conforms to ASTM6132 and ISO2808
  • Selectable display languages
The standard ultrasonic coating thickness gauges non-destructively measure the thickness of coatings on non-metal substrates including wood, plastic, composites, and more. They also measure the total thickness of a coating system.
509,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Display Range
Availability Pricing
DeFelsko PosiTector 200B1 Coating Thickness Gauge, Non-Metals, 0.5-40 mils WW-59784-02 Mfr # 200B1-E
COMPARE
LCD 0.5 to 40 mil
₹509,000.00INR / Each