This product is now discontinued, View Related Products

Danger: Respirator Required Signs

Protect your workers by clearly displaying areas requiring a respirator
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
DANGER (red): indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
Accuform MPPE159VS Safety Sign, Danger - Respirator Required, 7" x 10", Adhesive Vinyl WW-40711-93 Mfr # MPPE159VS
COMPARE
7 Vinyl 10
Call For Price
Accuform MPPE159VP Safety Sign, Danger - Respirator Required, 7" x 10", Plastic WW-40711-94 Mfr # MPPE159VP
COMPARE
7 Plastic 10
Call For Price
Accuform MPPE159VA Safety Sign, Danger - Respirator Required, 7" x 10", Aluminum WW-40711-95 Mfr # MPPE159VA
COMPARE
7 Aluminum 10
Call For Price
Accuform MPPA034VS Safety Sign, Danger - Respirator Required, 10" x 14", Adhesive Vinyl WW-40711-96 Mfr # MPPA034VS
COMPARE
10 Vinyl 14
Call For Price
Accuform MPPA034VP Safety Sign, Danger - Respirator Required, 10" x 14", Plastic WW-40711-97 Mfr # MPPA034VP
COMPARE
10 Plastic 14
Call For Price
Accuform MPPA034VA Safety Sign, Danger - Respirator Required, 10" x 14", Aluminum WW-40711-98 Mfr # MPPA034VA
COMPARE
10 Aluminum 14
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat