This product is now discontinued, View Related Products

Danger: Open Hole Signs

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.

DANGER (red): Indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
Accuform MSTF102VS Safety Sign, Danger - Open Hole, 7" X 10", Adhesive Vinyl WW-40710-69 Mfr # MSTF102VS
COMPARE
7 Adhesive Vinyl 10
Call For Price
Accuform MSTF102VP Safety Sign, Danger - Open Hole, 7" X 10", Plastic WW-40710-70 Mfr # MSTF102VP
COMPARE
7 Plastic 10
Call For Price
Accuform MSTF102VA Safety Sign, Danger - Open Hole, 7" X 10", Aluminum WW-40710-71 Mfr # MSTF102VA
COMPARE
7 Aluminum 10
Call For Price
Accuform MCSPD30VS Safety Sign, Danger - Open Hole, 10" X 14", Adhesive Vinyl WW-41014-69 Mfr # MCSPD30VS
COMPARE
10 Adhesive Vinyl 14
Call For Price
Accuform MCSPD30VP Safety Sign, Danger - Open Hole, 10" X 14", Plastic WW-41014-70 Mfr # MCSPD30VP
COMPARE
10 Plastic 14
Call For Price
Accuform MCSPD30VA Safety Sign, Danger - Open Hole, 10" X 14", Aluminum WW-41014-71 Mfr # MCSPD30VA
COMPARE
10 Aluminum 14
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat