This product is now discontinued, View Related Products

Danger: No Trespassing Signs

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
DANGER (red): Indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
Accuform MADM292VS Safety Sign, Danger - No Trespassing, 7" X 10", Adhesive Vinyl WW-40710-09 Mfr # MADM292VS
COMPARE
7 Adhesive Vinyl 10
Call For Price
Accuform MADM292VP Safety Sign, Danger - No Trespassing, 7" X 10", Plastic WW-40710-10 Mfr # MADM292VP
COMPARE
7 Plastic 10
Call For Price
Accuform MADM292VA Safety Sign, Danger - No Trespassing, 7" X 10", Aluminum WW-40710-11 Mfr # MADM292VA
COMPARE
7 Aluminum 10
Call For Price
Accuform MADM076VS Safety Sign, Danger - No Trespassing, 10" X 14", Adhesive Vinyl WW-41014-09 Mfr # MADM076VS
COMPARE
10 Adhesive Vinyl 14
Call For Price
Accuform MADM076VP Safety Sign, Danger - No Trespassing, 10" X 14", Plastic WW-41014-10 Mfr # MADM076VP
COMPARE
10 Plastic 14
Call For Price
Accuform MADM076VA Safety Sign, Danger - No Trespassing, 10" X 14", Aluminum WW-41014-11 Mfr # MADM076VA
COMPARE
10 Aluminum 14
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat