Cole-Parmer® Essentials Cylindrical HDPE Closed-Dome Tanks
FDA compliant
All tanks have a threaded screw closure with gasket that secuely fits the 6 1/4" fill well. Two mounting flats near the bottom, set 180°> apart, permit installation of a bulkhead fitting up to 2". Domed bottom drains fluid into narrow trough around base rim. High-density polyethylene tanks have a 2" bung with with cap on the top that permits access for tubing or piping. All tanks are FDA compliant.
67,100.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Volume (Gal) Volume (Liters)
Availability Pricing
COMPARE
HDPE 30 114
₹67,100.00INR / Each
COMPARE
HDPE 55 208
Call For Price
COMPARE
HDPE 100 379