Corning® Transwell® Sterile Permeable Supports
Essential tool for the study of both anchorage-dependent and independent cell lines
  • Provide an environment for cells that closely resembles their in vivo state
  • Treated for optimal cell attachment
  • Ideal for a variety of cell culture applications—chemotaxis, drug transport, tissue remodeling, and invasion studies
  • Supports come sterile and preloaded in cluster plates
  • Sterilized by gamma radiation
These permeable supports, also known as cell culture inserts, are designed to satisfy your diverse cell culture requirements. A suspended design prevents media wicking between the insert and the plate well. It also allows for co-culturing without damaging cells attached to the plate's growth surface. Choose either polycarbonate (PC) or Polyethylene Terephthalate (PET) membrane material. Polycarbonate membranes are translucent while PET membranes are clear, allowing for excellent visibility under phase contrast microscopy.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Cluster Plate Size Number Membrane Material Pore Size (µm)
Availability Pricing
Corning Transwell® Permeable Supports, Sterile, Polycarbonate Membrane, 0.4 µm, 6-Cluster Plate; 24/Cs WW-19480-00 Mfr # 3412
COMPARE
6 Polycarbonate 0.4
Call For Price
Corning Transwell® Permeable Supports, Sterile, Polycarbonate Membrane, 0.4 µm, 12-Cluster Plate; 48/Cs WW-19480-03 Mfr # 3401
COMPARE
12 Polycarbonate 0.4
Call For Price
Corning Transwell® Permeable Supports, Sterile, Polycarbonate Membrane, 0.4 µm, 24-Cluster Plate; 48/Cs WW-19480-07 Mfr # 3413
COMPARE
24 Polycarbonate 0.4
Call For Price
Corning Transwell® Permeable Supports, Sterile, Polycarbonate Membrane, 5 µm, 24-Cluster Plate; 48/Cs WW-19480-09 Mfr # 3421
COMPARE
24 Polycarbonate 5
Call For Price
Corning Transwell® Permeable Supports, Sterile, Polycarbonate Membrane, 8 µm, 24-Cluster Plate; 48/Cs WW-19480-10 Mfr # 3422
COMPARE
24 Polycarbonate 8
Call For Price
Corning Transwell® Permeable Supports, Sterile, PET Membrane, 0.4 µm, 6-Cluster Plate; 24/Cs WW-19480-13 Mfr # 3450
COMPARE
6 Polyethylene Terephthalate (PET) 0.4
Call For Price
Corning Transwell® Permeable Supports, Sterile, PET Membrane, 0.4 µm, 12-Cluster Plate; 48/Cs WW-19480-15 Mfr # 3460
COMPARE
12 Polyethylene Terephthalate (PET) 0.4
Call For Price
Corning Transwell® Permeable Supports, Sterile, PET Membrane, 0.4 µm, 24-Cluster Plate; 48/Cs WW-19480-17 Mfr # 3470
COMPARE
24 Polyethylene Terephthalate (PET) 0.4
Call For Price
Showing 1- 8 of 8