This product is now discontinued, View Related Products
Corning Tissue Culture Plate with Ultra-Low Attachment Surface
Unique hydrogel surface inhibits cell attachment
These culture plates and dishes have a unique covalently bonded hydrogel surface that is hydrophilc and neutrally charged. It minimizes cell attachment, protein absorption and enzyme activation. The surface is non-cytotoxic, biologically inert and nondegradable. Use these products to maintain cells in a suspended, unattached state, prevent stem cells from attachment-mediated differentiation, prevent anchorage-dependent cells from dividing, and reduce binding of attachment and serum proteins to the substrate.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Membrane Material Sterile Surface Treatment
Availability Pricing
Corning 3262 ultra low attachment surface culture dish, round dish, 100 mm dia WW-01835-22 Mfr # 3262
COMPARE
Hydrogel