Corning® ThermalTray™ Thermoconductive Platforms
Don't let your samples sinkDon't let your samples sink
  • Support containers and tube racks in liquid temperature-control media
  • Highly conductive alloy is compatible with liquid nitrogen
  • Sterilize by autoclave or dry heat
These thermally conductive trays support samples in water baths, melting ice, and liquid nitrogen, keeping the sample out of the liquid environment. Alloy construction is sturdy, stable, and ensures a uniform temperature across entire surface. Resistant to bleach, alcohol, other disinfectants, and laboratory detergents.
93,200.00 - ₹97,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Length (") Width (InchShort)
Availability Pricing
Corning SLP ThermalTray Thermoconductive Platform, slim, low profile; 1/ea WW-04392-73 Mfr # 432073
COMPARE
1 1/4 5 1/2
₹93,200.00INR / Each
Corning LP ThermalTray Thermoconductive Platform, low profile; 1/ea WW-04392-74 Mfr # 432074
COMPARE
2 5 1/2
₹97,000.00INR / Each