Corning IsoTips Filtered Pipette Tips
Prevents cross-contamination in critical application
  • Packaged sterile
  • RNase-/DNase-free
  • Nonpyrogenic
These IsoTips feature an inert, hydrophobic barrier which prevents aerosolized contaminants from coming in contact with pipettor shafts,. They are manufactured using Statistical Process Control (SPC) in their modeling operation to ensure premium quality. This process yields consistent critical dimensions such as tip straightness, length, and sealing ring location. Tips will properly attach to and eject from your pipettor everytime you use them. Ideal for preventing cross contamination is critical, such as DNA amplification and radioisotope handling.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL)
Availability Pricing
Corning 4807 IsoTip, 0.1 to 10 µL, G tip, sterile, 960/pk WW-24850-90 Mfr # 4807
COMPARE
10 0.1
Call For Price
Corning 4823 IsoTip Filtered Pipette Tip, sterile, 1-200 uL, 960/pk WW-24850-92 Mfr # 4823
COMPARE
200 1