This product is now discontinued, View Related Products

Corning® Gosselin™ Straight-Sided Hinged-Cap Containers, PP

Effortlessly collect liquid samples with one hand
  • Highly resistant to chemicals and breakage—all plastic
  • Molded graduation marks for precise container filling
  • Label with a pen or marker—convenient, frosted writing area
  • Sterile container to prevent contamination
These multipurpose, straight-sided bottles are ideal for handling liquid samples. The all-polypropylene construction offers excellent chemical resistance—perfect for storing diluted bases and acids. These containers are available in suitable volume sizes for milk sampling collection, or storage of small components.


Each container comes with an attached, self-standing cap—simply push down to close. Bottles arrive at your site inside a PE bag, then packed inside a box with label to show lot number and expiration date—traceable.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Label Type Special Features
Availability Pricing
Corning Gosselin FL52-02 Straight-Sided Hinged-Cap Containers, PP, 45 mL; 650/Cs WW-34551-18 Mfr # FL52-02
COMPARE
45 Frosted Sterile
Call For Price
Corning Gosselin FL60-04 Straight-Sided Hinged-Cap Containers, PP, 50 mL; 650/Cs WW-34551-19 Mfr # FL60-04
COMPARE
50 Frosted Sterile
Call For Price
Corning Gosselin FL100-05 Straight-Sided Hinged-Cap Containers, PP, 90 mL; 350/Cs WW-34551-20 Mfr # FL100-05
COMPARE
90 Frosted Sterile
Call For Price
Corning Gosselin FL300-04 Straight-Sided Hinged-Cap Containers, PP, 300 mL, 240/Cs WW-34551-27 Mfr # FL300-04
COMPARE
300 Not applicable Sterile
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat