Corning® Gosselin™ Sterile Snap-Cap Containers, PP
Economical and disposable sample containers for chemicals and electronic parts
  • Keep samples safe inside a leakproof PP snap-cap container
  • Highly resistant to chemicals and electricity
  • Extremely shockproof to easily take impacts around the workplace
  • Precise container filling with the help of the molded graduation marks
  • Convenient writing area on the container for labeling with a pen or marker
  • Available in sterile and nonsterile options
Graduated PP sample containers are an ideal choice for storing diluted bases and acids, and for applications where containers need to keep their shape after bending and flexing. As well as the storage of electronic parts that need electrical insulation. Each container comes with a PE snap cap for a leakproof seal.
Containers are protected from particle contamination during shipping. Containers arrive at your site enclosed in a PE bag packed inside a box with a label to show the lot number and expiration date for tracing purposes.
31,900.00INR / Case of 80
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bottle Color Capacity (mL) Description
Availability Pricing
Corning Gosselin PC1000-05 Sterile Snap-Cap Container, PP, 1000 mL, Natural; 80/Cs WW-34551-29 Mfr # PC1000-05
COMPARE
Natural 1000 Sterile Snap-Cap Container, PP, 1000 mL, Natural; 80/Cs
₹31,900.00INR / Case of 80