Corning® Gosselin™ Bi-Plate Petri Dishes, Polystyrene
Optimized for automation and double-bagged for security
  • Optimal flatness ensures constant depth of agar
  • Three vents for easy gas exchange
  • Class 100 manufacturing environment ensures aseptic production and packaging
Bi-plate petri dishes are designed for use with automatic filling equipment. Dishes are constructed of crystal polystyrene with the injection point located on the side of the dish bottom to optimize clarity even by image analyzing systems.
Dishes are available in aseptic and sterile versions and feature cleanroom packaging with two outer bags and bar code label. Aseptic products are manufactured in a controlled environment, without human intervention during the manufacturing process. Sterile products are sterilized using an irradiation method compliant with the requirements of ANSI/AAMI/ISO 11137 to achieve (SAL 10⁻⁶).
31,600.00 - ₹34,600.00INR / Case of 825
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sterile Description
Availability Pricing
Corning Gosselin Bi-Plate Petri Dish, Polystyrene, 100 x 15 mm, Sterile; 825/Cs WW-34552-05 Mfr # BI93D-102
COMPARE
Sterile Bi-Plate Petri Dish, Polystyrene, 100 x 15 mm, Sterile; 825/Cs
₹34,600.00INR / Case of 825
Corning Gosselin Bi-Plate Petri Dish, Polystyrene, 100 x 15 mm, Aseptic; 825/Cs WW-34552-06 Mfr # BI93D-101
COMPARE
Aseptic Bi-Plate Petri Dish, Polystyrene, 100 x 15 mm, Aseptic; 825/Cs
₹31,600.00INR / Case of 825