Corning® CoolCell® Filler Vials for CoolCell® Containers
No need to freeze a full load of samples
  • Ensure cell freezing rate consistency and uniform results
Use filler vials when you need to freeze less than a full load of samples to maintain a consistent cooling profile. To ensure cell freezing rate consistency and uniform results when using CoolCell containers, insert a CoolCell Filler Vial into empty wells when freezing less than a full load.
7,200.00INR / Pkg of 6
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Compatible With
Availability Pricing
Corning 432076 CoolCell® Filler Vials, 2 mL; 6/Pk WW-04392-76 Mfr # 432076
COMPARE
2 CoolCell LX, CoolCell FTS30 and CoolCell 5ml LX containers
₹7,200.00INR / Pkg of 6
Corning 432077 CoolCell® Filler Vials, 5 mL; 6/Pk WW-04392-77 Mfr # 432077
COMPARE
5 CoolCell 5ml LX containers
₹7,200.00INR / Pkg of 6