Corning® Corning® Cryogenic Vials, Sterile, Polypropylene
Safely and securely store your cryogenic samples
  • Supplied sterile and certified non-pyrogenic
  • Polypropylene construction withstands temperatures to -196°C
  • Silicone washers provide a secure seal
Vials feature a self-standing design with black graduations and larger marking spots for easier filling and sample identification. Choose from internal or external threaded vials for a wide range of applications.
Vial caps will hold Corning® color-coded inserts and will lock into Corning® cryogenic racks for single-handed operation. Cap inserts and self-locking racks are sold separately.
7,000.00 - ₹7,500.00INR / Pkg of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Thread Tube Bottom Shape Volume (mL)
Availability Pricing
Corning Cryogenic Vials, Sterile, Polypropylene, Self-Standing with Round Bottom, External Thread, 2 mL; 50/Pk WW-44351-02 Mfr # 430659
COMPARE
External Self-Standing with Round Bottom 2
₹7,000.00INR / Pkg of 50
Corning Cryogenic Vials, Sterile, Polypropylene, Self-Standing with Round Bottom, Internal Thread, 2 mL; 50/Pk WW-44351-12 Mfr # 430488
COMPARE
Internal Self-Standing with Round Bottom 2
₹7,500.00INR / Pkg of 50