Corning Chemistry Kits
Enables standard laboratory assemblies to be constructed with ease and efficiency!
Kits include a variety of sizes for conducting experiments at various volume stages. Manufactured from glass, the round boiling flasks wall thickness is controlled to provide the proper balance between thermal expansion and mechanical strength for a uniform heating surface. Use for applications such as distillation, reaction with gas evolution, preparation or recovery, simple reflux; reflux with addition, fractional distillation and vacuum distillation. Standard joint sizes make assembly easy.
180,000.00 - ₹232,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Joint Size Number Description
Availability Pricing
Corning 6949E Chemistry Kit with 19/22 joint sizes WW-67972-30 Mfr # 6949E
COMPARE
19/22 Chemistry Kit with 19/22 joint sizes
₹180,000.00INR / Each
Corning 6949 Chemistry Kit with 24/40 joint size WW-67972-40 Mfr # 6949
COMPARE
24/40 Chemistry Kit with 24/40 joint size
₹232,000.00INR / Each