Corning® Sterile Cell Culture Flasks, Polystyrene
Get better traceability
  • Virgin polystyrene provides excellent optical quality
  • Gamma radiated, certified nonpyrogenic, and nontoxic
These sterile culture flasks have individually printed lot numbers for product traceability and feature molded-on graduations. Flasks are constructed of durable polystyrene and have an HDPE cap. Choose from a range of cap types, neck styles, flask shapes, and surface treatment options for many cell culture applications.
Cap Type options include a plug seal for a liquid and gas tight seal, phenolic sold flat top caps, and vented caps which contain a 0.2 μm nonwetting (hydrophobic) membrane sealed to the cap allowing gas exchange.
Neck Style options include canted and angled. Canted necks feature a ramp from the bottom to the neck allowing for easy access to growth surface for pipetting or scraping. Angled necks utilize the entire bottom area for cell growth, saving space and reducing the medium sloshing into the neck.
Flask Shapes available are rectangular and U-shaped. Rectangular flasks feature sharp corners for enhances stability. U-shaped flasks have rounded shoulders for an easier grip and better access when removing or tightening the cap.
Surface Treatment options are the standard tissue culture surface treatment for optimal cell attachment and ultra-low attachment which features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Cap Type Culture Area (cm²) Surface Treatment
Availability Pricing
Corning Cell Culture Flask, Polystyrene, Sterile, Tissue Culture Treated, Plug Seal, 25 cm2; 500/Cs WW-01936-00 Mfr # 430168
COMPARE
Plug Seal 25 Tissue Culture
Call For Price
Corning Cell Culture Flask, Polystyrene, Sterile, Tissue Culture Treated, Vent Cap, 25 cm2; 200/Cs WW-01936-02 Mfr # 430639
COMPARE
Vented 25 Tissue Culture
Call For Price
Corning Cell Culture Flask, Polystyrene, Sterile, Tissue Culture Treated, Plug Seal, 150 cm2; 50/Cs WW-01936-20 Mfr # 430823
COMPARE
Plug Seal 150 Tissue Culture
Call For Price
Corning Cell Culture Flask, Polystyrene, Sterile, Tissue Culture Treated, Vent Cap, 150 cm2; 50/Cs WW-01936-22 Mfr # 430825
COMPARE
Vented 150 Tissue Culture
Call For Price
Corning Cell Culture Flask, Polystyrene, Sterile, Tissue Culture Treated, Vent Cap, 175 cm2; 50/Cs WW-01936-32 Mfr # 431080
COMPARE
Vented 175 Tissue Culture
Call For Price
Corning Cell Culture Flask, Polystyrene, Sterile, Tissue Culture Treated, Phenolic Cap, 25 cm2; 500/Cs WW-01936-50 Mfr # 430372
COMPARE
Phenolic 25 Tissue Culture
Call For Price
Corning Cell Culture Flask, Polystyrene, Sterile, Tissue Culture Treated, Plug Seal, 175 cm2; 50/Cs WW-01936-54 Mfr # 431079
COMPARE
Plug Seal 175 Tissue Culture
Call For Price
Showing 1- 7 of 7