Corning® 96-Well Sterile Microplate Lids, Polystyrene
Designed to reduce contamination
  • Constructed of rigid polystyrene
  • Sterilized by gamma radiation
The clear polystyrene (PS) lids are well suited for the Corning ® 96-well plates. Lids feature a corner-notch design with condensation rings to reduce contamination.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Packaging Plate Color Sterile
Availability Pricing
Corning 96-well Microplate Lid, Low Evaporation Lid with Corner Notch, Sterile, Polystyrene; 100/Cs WW-01730-01 Mfr # 3930
COMPARE
1 lid per pack and 100 lids per case Clear Yes
Call For Price
Corning 96-well Microplate Lid, Low Evaporation Lid with Corner Notch, Sterile, Polystyrene; 50/Cs WW-01730-02 Mfr # 3931
COMPARE
25 lids per pack and 50 lids per case Clear Yes